missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Thienyl)pyridine 95.0+%, TCI America™

Catalog No. T11515G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T11515G Supplier TCI America Supplier No. T11515G
Only null left

Chemical Identifiers

CAS 21298-55-5
Molecular Formula C9H7NS
Molecular Weight (g/mol) 161.222
MDL Number MFCD00006214
InChI Key MINKMXOCFYGGKA-UHFFFAOYSA-N
Synonym 3-(2-Pyridyl)thiophene
PubChem CID 88862
IUPAC Name 2-thiophen-3-ylpyridine
SMILES C1=CC=NC(=C1)C2=CSC=C2

Specifications

Quantity 5 g
Formula Weight 161.22
Percent Purity ≥95.0% (GC)
Physical Form Lumps
Chemical Name or Material 2-(3-Thienyl)pyridine