missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Pyridyl)benzimidazole, 97%

Catalog No. AAH5178514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5178514 25 g
AAH5178506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5178514 Supplier Thermo Scientific Chemicals Supplier No. H5178514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1137-67-3 | C12H9N3 | 195.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1137-67-3
Molecular Formula C12H9N3
Molecular Weight (g/mol) 195.23
MDL Number MFCD00022665
InChI Key BOUOQESVDURNSB-UHFFFAOYSA-N
Synonym 2-pyridin-3-yl-1h-benzo d imidazole, 2-3-pyridyl benzimidazole, 2-3-pyridyl-1h-benzimidazole, 2-pyridin-3-yl-1h-1,3-benzodiazole, 2-3-pyridinyl-1h-benzimidazole, 2-pyridin-3-yl-1h-benzoimidazole, chembl83103, 2-pyridin-3-yl-1h-benzimidazole, 1h-benzimidazole, 2-3-pyridinyl, ldha inhibitor, 10
PubChem CID 247635
IUPAC Name 2-pyridin-3-yl-1H-benzimidazole
SMILES N1C2=CC=CC=C2N=C1C1=CC=CN=C1

Specifications

Melting Point 254°C to 260°C
Odor Odorless
Quantity 25 g
Formula Weight 195.22
Percent Purity 97%
Chemical Name or Material 2-(3-Pyridyl)benzimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only