missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Pyridyl)benzimidazole-6-carboxylic acid, 97%

Catalog No. p-7036321
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Catalog No. Quantity
AAH54593MD 250 mg
AAH5459303 1 g
AAH5459306 5 g
3 options

Catalog No. AAH54593MD

Supplier: Thermo Scientific Chemicals H54593MD

Only null left

CAS: 312496-16-5 | C13H9N3O2 | 239.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 312496-16-5
Molecular Formula C13H9N3O2
Molecular Weight (g/mol) 239.234
MDL Number MFCD01414693
InChI Key DHRPCXYIVJXXJR-UHFFFAOYSA-N
Synonym 1h-benzimidazole-6-carboxylicacid, 2-3-pyridinyl, 2-pyridin-3-yl-3h-1,3-benzodiazole-5-carboxylic acid, 2-pyridin-3-yl-3h-benzoimidazole-5-carboxylic acid, 2-3-pyridyl benzimidazole-6-carboxylic acid, 6-carboxy-2-pyridin-3-yl-1h-benzimidazole, 2-pyridin-3-yl-1h-benzoimidazole-5-carboxylic acid, 2-3-pyridinyl-1h-benzoimidazole-5-carboxylic acid, 2-pyridin-3-yl-1h-benzimidazole-5-carboxylic acid, 2-pyridin-3-yl-3h-benzimidazole-5-carboxylic acid
PubChem CID 2771805
IUPAC Name 2-pyridin-3-yl-3H-benzimidazole-5-carboxylic acid
SMILES C1=CC(=CN=C1)C2=NC3=C(N2)C=C(C=C3)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 239.23
Percent Purity 97%
Chemical Name or Material 2-(3-Pyridyl)benzimidazole-6-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only