missing translation for 'onlineSavingsMsg'
Learn More

2,3-Pyridinedicarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7134669
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P055025G 25 g
P0550500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P055025G Supplier TCI America Supplier No. P055025G
Only null left

Chemical Identifiers

CAS 89-00-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00006295
InChI Key GJAWHXHKYYXBSV-UHFFFAOYSA-N
Synonym quinolinic acid, 2,3-pyridinedicarboxylic acid, quinolinate, pyridine-2,3-dicarboxylate, unii-f6f0hk1urn, 2,3-pyridinedicarboxylicacid, pyridine-2,3-carboxylate, f6f0hk1urn, pyridin-2,3-dicarbonsaeure, 2,3-pyridinedicarboxylate
PubChem CID 1066
ChEBI CHEBI:16675
IUPAC Name pyridine-2,3-dicarboxylic acid
SMILES C1=CC(=C(N=C1)C(=O)O)C(=O)O

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 25 g
Formula Weight 167.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Pyridinedicarboxylic Acid
EINECSNumber (5)-5505
RTECSNumber US7967250
TSCA Yes