missing translation for 'onlineSavingsMsg'
Learn More

2,3-Pyridinedicarboxylic acid, 99%

Catalog No. p-189281
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131850250 25 g Plastic bottle
AC131851000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC131850250 Supplier Thermo Scientific Chemicals Supplier No. 131850250
Only null left

CAS: 89-00-9 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-00-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00006295
InChI Key GJAWHXHKYYXBSV-UHFFFAOYSA-N
Synonym quinolinic acid, 2,3-pyridinedicarboxylic acid, quinolinate, pyridine-2,3-dicarboxylate, unii-f6f0hk1urn, 2,3-pyridinedicarboxylicacid, pyridine-2,3-carboxylate, f6f0hk1urn, pyridin-2,3-dicarbonsaeure, 2,3-pyridinedicarboxylate
PubChem CID 1066
ChEBI CHEBI:16675
IUPAC Name pyridine-2,3-dicarboxylic acid
SMILES C1=CC(=C(N=C1)C(=O)O)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 188°C to 190°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 25 g
Beilstein 22, 150
Merck Index 15, 8190
Solubility Information Solubility in water: 0.55%. Other solubilities: soluble in alkalies,slightly soluble in alcohol,practically insoluble in ether and benzene
Formula Weight 167.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Pyridinedicarboxylic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 201-874-8
RTECSNumber US7967250
TSCA TSCA

RUO – Research Use Only