missing translation for 'onlineSavingsMsg'
Learn More

2,3-Pyrazinedicarboxylic anhydride, 97%

Catalog No. AC397130050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397130050 5 g Glass bottle
AC397130250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397130050 Supplier Thermo Scientific Chemicals Supplier No. 397130050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4744-50-7 | C6H2N2O3 | 150.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4744-50-7
Molecular Formula C6H2N2O3
Molecular Weight (g/mol) 150.09
MDL Number MFCD00179418
InChI Key AWJWCTOOIBYHON-UHFFFAOYSA-N
Synonym furo 3,4-b pyrazine-5,7-dione, 2,3-pyrazinedicarboxylic anhydride, 2,3-pyrazinecarboxylic anhydride, pyrazine-2,3-dicarboxylic anhydride, 5h,7h-furo 3,4-b pyrazine-5,7-dione, furano 3,4-b pyrazine-5,7-dione, pubchem17535, 2,3-pyrazinedicarboxylic anhydride treated bsa
PubChem CID 78482
IUPAC Name 5H,7H-furo[3,4-b]pyrazine-5,7-dione
SMILES O=C1OC(=O)C2=NC=CN=C12

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 210°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 150.09
Percent Purity 97%
Chemical Name or Material 2, 3-Pyrazinedicarboxylic anhydride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
EINECSNumber 225-260-4

RUO – Research Use Only