missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Phenylpropyl)pyridine, 99%

Catalog No. p-7026483
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2249503 1 g
AAB2249506 5 g
AAB2249514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2249503 Supplier Thermo Scientific Chemicals Supplier No. B2249503
Only null left

CAS: 2110-18-1 | C14H15N | 197.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2110-18-1
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.28
MDL Number MFCD00191237
InChI Key JJJPNTQYUJPWGQ-UHFFFAOYSA-N
Synonym 2-3-phenylpropyl pyridine, pyridine, 2-3-phenylpropyl, unii-9f0bfm744f, alpha-3-phenylpropyl pyridine, 2-phenylpropylpyridine, cortex pyridine, phenylpropylpyridine, 3-phenylpropyl pyridine, acmc-1cs6o, dsstox_cid_22356
PubChem CID 459494
IUPAC Name 2-(3-phenylpropyl)pyridine
SMILES C(CC1=CC=CC=C1)CC1=CC=CC=N1

Specifications

Density 1.017
Boiling Point 142°C to 143°C (1 mmHg)
Flash Point 100°C (212°F)
Quantity 1 g
Formula Weight 197.28
Percent Purity 99%
Chemical Name or Material 2-(3-Phenylpropyl)pyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 218-300-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only