missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Pentyl)pyridine 95.0+%, TCI America™

Catalog No. P096925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P096925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P096925ML Supplier TCI America Supplier No. P096925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7399-50-0
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00014631
InChI Key FTYAXYWEDPWJCJ-UHFFFAOYSA-N
Synonym 3-(2-Pyridyl)pentane
PubChem CID 81887
IUPAC Name 2-(pentan-3-yl)pyridine
SMILES CCC(CC)C1=CC=CC=N1

Specifications

Quantity 25 mL
Formula Weight 149.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3-Pentyl)pyridine