missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',3'-O-Isopropylideneguanosine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 296°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Physical Form | Crystal-Powder at 20°C |
| Formula Weight | 323.31 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Chemical Name or Material | 2′,3′-O-Isopropylideneguanosine |
Safety and Handling
| TSCA | No |