missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-2,3-O-Isopropylidene-L-threitol 97.0+%, TCI America™
$136.08 - $393.32
Chemical Identifiers
| CAS | 50622-09-8 |
|---|---|
| Molecular Formula | C7H14O4 |
| Molecular Weight (g/mol) | 162.185 |
| MDL Number | MFCD00063761 |
| InChI Key | INVRLGIKFANLFP-WDSKDSINSA-N |
| Synonym | +-2,3-o-isopropylidene-l-threitol, 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-diyl dimethanol, 2,3-o-isopropylidene-l-threitol, 4s,5s-5-hydroxymethyl-2,2-dimethyl-1,3-dioxolan-4-yl methanol, +-2,3,o-isopropylidene-l-threitol, pubchem19000, 2,3-o-isoproylidene-l-threitol, 1,3-dioxolane-4,5-dimethanol, 2,2-dimethyl-, 4s,5s, 2-o,3-o-isopropylidene-l-threitol, oi-2,3-o-isopropylidene-l-threitol |
| PubChem CID | 785242 |
| IUPAC Name | [(4S,5S)-5-(hydroxymethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol |
| SMILES | CC1(OC(C(O1)CO)CO)C |
Chemical Identifiers
| 50622-09-8 | |
| 162.185 | |
| INVRLGIKFANLFP-WDSKDSINSA-N | |
| 785242 | |
| CC1(OC(C(O1)CO)CO)C |
| C7H14O4 | |
| MFCD00063761 | |
| +-2,3-o-isopropylidene-l-threitol, 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-diyl dimethanol, 2,3-o-isopropylidene-l-threitol, 4s,5s-5-hydroxymethyl-2,2-dimethyl-1,3-dioxolan-4-yl methanol, +-2,3,o-isopropylidene-l-threitol, pubchem19000, 2,3-o-isoproylidene-l-threitol, 1,3-dioxolane-4,5-dimethanol, 2,2-dimethyl-, 4s,5s, 2-o,3-o-isopropylidene-l-threitol, oi-2,3-o-isopropylidene-l-threitol | |
| [(4S,5S)-5-(hydroxymethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol |
Specifications
| 50622-09-8 | |
| White-Yellow | |
| MFCD00063761 | |
| +-2,3-o-isopropylidene-l-threitol, 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-diyl dimethanol, 2,3-o-isopropylidene-l-threitol, 4s,5s-5-hydroxymethyl-2,2-dimethyl-1,3-dioxolan-4-yl methanol, +-2,3,o-isopropylidene-l-threitol, pubchem19000, 2,3-o-isoproylidene-l-threitol, 1,3-dioxolane-4,5-dimethanol, 2,2-dimethyl-, 4s,5s, 2-o,3-o-isopropylidene-l-threitol, oi-2,3-o-isopropylidene-l-threitol | |
| CC1(OC(C(O1)CO)CO)C | |
| 162.185 | |
| 162.19 | |
| Crystalline Lumps |
| 46°C | |
| C7H14O4 | |
| 1 g | |
| INVRLGIKFANLFP-WDSKDSINSA-N | |
| [(4S,5S)-5-(hydroxymethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol | |
| 785242 | |
| ≥97.0% (GC) | |
| (+)-2,3-O-Isopropylidene-L-threitol |
Safety and Handling
TSCA : No