missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Naphthalenedicarboxylic Anhydride 95.0+%, TCI America™
$339.25 - $1366.35
Chemical Identifiers
| CAS | 716-39-2 |
|---|---|
| Molecular Formula | C12H6O3 |
| Molecular Weight (g/mol) | 198.177 |
| MDL Number | MFCD00059091 |
| InChI Key | IZJDCINIYIMFGX-UHFFFAOYSA-N |
| PubChem CID | 69743 |
| IUPAC Name | benzo[f][2]benzofuran-1,3-dione |
| SMILES | C1=CC=C2C=C3C(=CC2=C1)C(=O)OC3=O |
Chemical Identifiers
| 716-39-2 | |
| 198.177 | |
| IZJDCINIYIMFGX-UHFFFAOYSA-N | |
| benzo[f][2]benzofuran-1,3-dione |
| C12H6O3 | |
| MFCD00059091 | |
| 69743 | |
| C1=CC=C2C=C3C(=CC2=C1)C(=O)OC3=O |
Specifications
| 716-39-2 | |
| 380°C | |
| MFCD00059091 | |
| IZJDCINIYIMFGX-UHFFFAOYSA-N | |
| benzo[f][2]benzofuran-1,3-dione | |
| 69743 | |
| ≥95.0% (GC,T) | |
| 2,3-Naphthalenedicarboxylic Anhydride |
| 248°C | |
| C12H6O3 | |
| 25 g | |
| C1=CC=C2C=C3C(=CC2=C1)C(=O)OC3=O | |
| 198.177 | |
| 198.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes