missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Naphthalenedicarboxylic Anhydride 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 248°C |
| Boiling Point | 380°C |
| Quantity | 25 g |
| Formula Weight | 198.18 |
| Percent Purity | ≥95.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,3-Naphthalenedicarboxylic Anhydride |
Safety and Handling
| TSCA | Yes |