missing translation for 'onlineSavingsMsg'
Learn More

2,3-Lutidine, 99%

Catalog No. AAL0369714
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0369714 25 g
AAL0369722 100 g
AAL0369736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0369714 Supplier Thermo Scientific Chemicals Supplier No. L0369714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 583-61-9 | C7H9N | 107.156 g/mol

2,3-Lutidine is a lansoprazole intermediates. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Lutidine is a lansoprazole intermediates. It is used as pharmaceutical intermediate.

Solubility
Soluble in water 95 gl/ @ 20˚C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 583-61-9
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00009605
InChI Key HPYNZHMRTTWQTB-UHFFFAOYSA-N
Synonym 2,3-lutidine, pyridine, 2,3-dimethyl, pyridine, dimethyl, dimethylpyridine, unii-5q0f649h6g, 2,3-dimethyl-pyridine, 2-3-lutidine, pyridine,3-dimethyl, 2,3-d1methylpyridine, pubchem20520
PubChem CID 11420
IUPAC Name 2,3-dimethylpyridine
SMILES CC1=C(N=CC=C1)C

Specifications

Melting Point -17°C
Density 0.948
Boiling Point 160°C to 161°C
Flash Point 50°C (122°F)
Odor Unpleasant
Refractive Index 1.508
Quantity 25 g
UN Number UN2929
Beilstein 106418
Solubility Information Soluble in water 95 gl/ @ 20?C.
Formula Weight 107.16
Percent Purity 99%
Chemical Name or Material 2,3-Lutidine
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 209-514-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only