missing translation for 'onlineSavingsMsg'
Learn More

2,3-Lutidine, 98+%

Catalog No. AC268761000
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC268761000 100 mL
AC268760250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC268761000 Supplier Thermo Scientific Chemicals Supplier No. 268761000
Only null left

CAS: 583-61-9 | C7H9N | 107.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 583-61-9
MDL Number MFCD00009605
InChI Key HPYNZHMRTTWQTB-UHFFFAOYSA-N
Synonym 2,3-lutidine, pyridine, 2,3-dimethyl, pyridine, dimethyl, dimethylpyridine, unii-5q0f649h6g, 2,3-dimethyl-pyridine, 2-3-lutidine, pyridine,3-dimethyl, 2,3-d1methylpyridine, pubchem20520
PubChem CID 11420
IUPAC Name 2,3-dimethylpyridine
SMILES CC1=C(N=CC=C1)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -17.0°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 161.0°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 0.7% max. 2-Ethylpiridine (GC)
Packaging Glass bottle
Molecular Formula C7H9N
Refractive Index 1.5062 to 1.5082
Quantity 100 mL
Beilstein 20, 243
Specific Gravity 0.95
Solubility Information Solubility in water: 95g/L (26°C). Other solubilities: soluble in methanol and ether,miscible with dmf and thf
Molecular Weight (g/mol) 107.16
Formula Weight 107.16
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2, 3-Lutidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-514-1

RUO – Research Use Only