missing translation for 'onlineSavingsMsg'
Learn More

2,3-Hexanedione, 94%

Catalog No. AAL0466914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0466914 25 g
AAL0466906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0466914 Supplier Thermo Scientific Chemicals Supplier No. L0466914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3848-24-6 | C6H10O2 | 114.14 g/mol

2,3-Hexanedione is used to evaluate the influence of xanthan concentration on the release of aroma compounds in xanthan-thickened food model systems. It reacts with ethylenediamine to yield macrocyclic tetradentate 12-membered nitrogen donor (N4) ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Hexanedione is used to evaluate the influence of xanthan concentration on the release of aroma compounds in xanthan-thickened food model systems. It reacts with ethylenediamine to yield macrocyclic tetradentate 12-membered nitrogen donor (N4) ligand.

Solubility
Partly miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3848-24-6
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00009398
InChI Key MWVFCEVNXHTDNF-UHFFFAOYSA-N
Synonym 2,3-hexanedione, acetylbutyryl, acetyl butyryl, methyl propyl diketone, butyryl acetyl, hexanedione, unii-559anr3nvs, 2,3-hexandione, fema no. 2558, 559anr3nvs
PubChem CID 19707
ChEBI CHEBI:87583
IUPAC Name hexane-2,3-dione
SMILES CCCC(=O)C(C)=O

Specifications

Melting Point -30°C
Density 0.937
Boiling Point 127°C to 128°C
Flash Point 28°C (82°F)
Odor Characteristic
Refractive Index 1.412
Quantity 25 g
UN Number UN1224
Beilstein 1699896
Solubility Information Partly miscible in water.
Formula Weight 114.15
Percent Purity 94%
Chemical Name or Material 2,3-Hexanedione
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 223-350-8
RTECSNumber MO3140000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only