missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(3-Fluorophenyl)ethylamine 98.0+%, TCI America™
$160.30 - $637.51
Chemical Identifiers
| CAS | 404-70-6 |
|---|---|
| Molecular Formula | C8H11FN |
| Molecular Weight (g/mol) | 140.18 |
| MDL Number | MFCD00075376 |
| InChI Key | AUCVZEYHEFAWHO-UHFFFAOYSA-O |
| Synonym | 3-fluorophenethylamine, 2-3-fluorophenyl ethylamine, 2-3-fluoro-phenyl-ethylamine, 2-3-fluorophenyl ethanamine, 2-3-fluorophenyl ethan-1-amine, benzeneethanamine, 3-fluoro, m-fluorophenethylamine, 3-flourophenethylamine, m-fluorophenylethylamine, 3-fluoro phenethylamine |
| PubChem CID | 533928 |
| IUPAC Name | 2-(3-fluorophenyl)ethan-1-aminium |
| SMILES | [NH3+]CCC1=CC=CC(F)=C1 |
Chemical Identifiers
| 404-70-6 | |
| 140.18 | |
| AUCVZEYHEFAWHO-UHFFFAOYSA-O | |
| 533928 | |
| [NH3+]CCC1=CC=CC(F)=C1 |
| C8H11FN | |
| MFCD00075376 | |
| 3-fluorophenethylamine, 2-3-fluorophenyl ethylamine, 2-3-fluoro-phenyl-ethylamine, 2-3-fluorophenyl ethanamine, 2-3-fluorophenyl ethan-1-amine, benzeneethanamine, 3-fluoro, m-fluorophenethylamine, 3-flourophenethylamine, m-fluorophenylethylamine, 3-fluoro phenethylamine | |
| 2-(3-fluorophenyl)ethan-1-aminium |
Specifications
| 404-70-6 | |
| 87°C | |
| MFCD00075376 | |
| 2735 | |
| AUCVZEYHEFAWHO-UHFFFAOYSA-O | |
| 2-(3-fluorophenyl)ethan-1-aminium | |
| 533928 | |
| ≥98.0% (GC,T) | |
| 2-(3-Fluorophenyl)ethylamine |
| Yellow | |
| C8H11FN | |
| 5 g | |
| 3-fluorophenethylamine, 2-3-fluorophenyl ethylamine, 2-3-fluoro-phenyl-ethylamine, 2-3-fluorophenyl ethanamine, 2-3-fluorophenyl ethan-1-amine, benzeneethanamine, 3-fluoro, m-fluorophenethylamine, 3-flourophenethylamine, m-fluorophenylethylamine, 3-fluoro phenethylamine | |
| [NH3+]CCC1=CC=CC(F)=C1 | |
| 140.18 | |
| 139.17 | |
| Liquid |
Safety and Handling
TSCA : No