missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Fluorophenyl)ethylamine 98.0+%, TCI America™

Catalog No. F09305G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09305G 5 g
F093025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09305G Supplier TCI America Supplier No. F09305G

Chemical Identifiers

CAS 404-70-6
Molecular Formula C8H11FN
Molecular Weight (g/mol) 140.18
MDL Number MFCD00075376
InChI Key AUCVZEYHEFAWHO-UHFFFAOYSA-O
Synonym 3-fluorophenethylamine, 2-3-fluorophenyl ethylamine, 2-3-fluoro-phenyl-ethylamine, 2-3-fluorophenyl ethanamine, 2-3-fluorophenyl ethan-1-amine, benzeneethanamine, 3-fluoro, m-fluorophenethylamine, 3-flourophenethylamine, m-fluorophenylethylamine, 3-fluoro phenethylamine
PubChem CID 533928
IUPAC Name 2-(3-fluorophenyl)ethan-1-aminium
SMILES [NH3+]CCC1=CC=CC(F)=C1

Specifications

Color Yellow
Boiling Point 87°C
Quantity 5 g
UN Number 2735
Formula Weight 139.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(3-Fluorophenyl)ethylamine