missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diphenylpyrazine, 98%

Catalog No. AAA1470003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1470003 1 g
AAA1470006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1470003 Supplier Thermo Scientific Chemicals Supplier No. A1470003

CAS: 1588-89-2 | C16H12N2 | 232.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1588-89-2
Molecular Formula C16H12N2
Molecular Weight (g/mol) 232.286
MDL Number MFCD00053000
InChI Key PTZIVVDMBCVSMR-UHFFFAOYSA-N
Synonym 2,3-diphenyl-pyrazine, chembl48222, bis-phenyl pyrazine, maybridge1_007492, acmc-1bu2l
PubChem CID 243907
IUPAC Name 2,3-diphenylpyrazine
SMILES C1=CC=C(C=C1)C2=NC=CN=C2C3=CC=CC=C3

Specifications

Melting Point 119°C to 122°C
Odor Odorless
Quantity 1 g
Formula Weight 232.29
Percent Purity 98%
Chemical Name or Material 2,3-Diphenylpyrazine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only