missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diphenylmaleic Anhydride 95.0+%, TCI America™

Catalog No. p-7128011
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39431G 1 g
D39435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39431G Supplier TCI America Supplier No. D39431G
Only null left

Chemical Identifiers

CAS 4808-48-4
Molecular Formula C16H10O3
Molecular Weight (g/mol) 250.253
MDL Number MFCD00005521
InChI Key OUJCFCNZIUTYBH-UHFFFAOYSA-N
Synonym 2,3-diphenylmaleic anhydride, diphenylmaleic anhydride, 3,4-diphenyl-2,5-furandione, diphenylfuran-2,5-dione, 2,3-diphenylmaleicanhydride, 2,5-furandione, 3,4-diphenyl, acmc-1asck, 2, 3,4-diphenyl, diphenyl-maleic anhydride, 2,5-furandione,3,4-diphenyl
PubChem CID 78530
IUPAC Name 3,4-diphenylfuran-2,5-dione
SMILES C1=CC=C(C=C1)C2=C(C(=O)OC2=O)C3=CC=CC=C3

Specifications

Melting Point 160°C
Color Yellow
Boiling Point 236°C
Quantity 1 g
Formula Weight 250.25
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diphenylmaleic Anhydride