missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diphenylmaleic anhydride, 98%

Catalog No. p-7046661
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1561203 1 g
AAL1561206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1561203 Supplier Thermo Scientific Chemicals Supplier No. L1561203
Only null left

CAS: 4808-48-4 | C16H10O3 | 250.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4808-48-4
Molecular Formula C16H10O3
Molecular Weight (g/mol) 250.253
MDL Number MFCD00005521
InChI Key OUJCFCNZIUTYBH-UHFFFAOYSA-N
Synonym 2,3-diphenylmaleic anhydride, diphenylmaleic anhydride, 3,4-diphenyl-2,5-furandione, diphenylfuran-2,5-dione, 2,3-diphenylmaleicanhydride, 2,5-furandione, 3,4-diphenyl, acmc-1asck, 2, 3,4-diphenyl, diphenyl-maleic anhydride, 2,5-furandione,3,4-diphenyl
PubChem CID 78530
IUPAC Name 3,4-diphenylfuran-2,5-dione
SMILES C1=CC=C(C=C1)C2=C(C(=O)OC2=O)C3=CC=CC=C3

Specifications

Melting Point 157°C to 160°C
Boiling Point 236°C (15 mmHg)
Quantity 1 g
Beilstein 191180
Sensitivity Moisture sensitive
Formula Weight 250.26
Percent Purity 98%
Chemical Name or Material 2,3-Diphenylmaleic anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-370-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only