missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diphenyl-5-(4-methoxyphenyl)tetrazolium Chloride 80.0+%, TCI America™

Catalog No. b0339100mg
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
B0339100MG 100 mg
1 options

Catalog No. B0339100MG

Supplier: TCI America B0339100MG

Only null left

Chemical Identifiers

CAS 10560-45-9
Molecular Formula C20H17ClN4O
MDL Number MFCD00060032

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 364.83
Percent Purity ≥80.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diphenyl-5-(4-methoxyphenyl)tetrazolium Chloride