missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylpyrazine, 99%

Catalog No. AAA1262809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1262809 10 g
AAA1262818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1262809 Supplier Thermo Scientific Chemicals Supplier No. A1262809
Only null left

CAS: 5910-89-4 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5910-89-4
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006144
InChI Key OXQOBQJCDNLAPO-UHFFFAOYSA-N
Synonym pyrazine, 2,3-dimethyl, 2,3-dimethyl-1,4-diazine, 2,3-dimethyl-pyrazine, pyrazine, dimethyl, 2,3-dimethyl pyrazine, ccris 2928, unii-whf7883d0v, fema no. 3271, natural, dimethylpyrazine
PubChem CID 22201
IUPAC Name 2,3-dimethylpyrazine
SMILES CC1=NC=CN=C1C

Specifications

Melting Point -12°C
Density 1.022
Boiling Point 156°C
Flash Point 54°C (129°F)
Odor Pleasant
Refractive Index 1.507
Quantity 10 g
UN Number UN1993
Beilstein 107908
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2,3-Dimethylpyrazine
Show More Show Less
Hazard Category H226-H302-H315-H319
Hazard Statement GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 227-630-0
RTECSNumber UQ2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only