Learn More
2,3-Dimethylnaphthalene, 97%
CAS: 581-40-8 | C12H12 | 156.228 g/mol
Supplier: Thermo Scientific Chemicals L0838606
Description
It is used in the identification of individual aromatic hydrocarbons in kerosene fraction (b.p. 150-250°).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the identification of individual aromatic hydrocarbons in kerosene fraction (b.p. 150-250°).
Solubility
Solubility in toluene (almost transparency). Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 2,3-Dimethylnaphthalene | |
| 103°C to 106°C | |
| C12H12 | |
| 5 g | |
| guajen, naphthalene, 2,3-dimethyl, unii-35dj6se17o, 2,3-dmn, 2,3-dimethyl naphthalene, acmc-1ao3c, 2,3-dimethylnaphthalene, naphthalene, 2,3-dimethyl-8ci 9ci | |
| CC1=CC2=CC=CC=C2C=C1C | |
| 156.228 | |
| CHEBI:48615 | |
| 97% |
| 581-40-8 | |
| 265°C to 268°C | |
| MFCD00004119 | |
| 1852621 | |
| WWGUMAYGTYQSGA-UHFFFAOYSA-N | |
| 2,3-dimethylnaphthalene | |
| 11386 | |
| 156.23 |
Chemical Identifiers
| 581-40-8 | |
| 156.228 | |
| WWGUMAYGTYQSGA-UHFFFAOYSA-N | |
| 11386 | |
| 2,3-dimethylnaphthalene |
| C12H12 | |
| MFCD00004119 | |
| guajen, naphthalene, 2,3-dimethyl, unii-35dj6se17o, 2,3-dmn, 2,3-dimethyl naphthalene, acmc-1ao3c, 2,3-dimethylnaphthalene, naphthalene, 2,3-dimethyl-8ci 9ci | |
| CHEBI:48615 | |
| CC1=CC2=CC=CC=C2C=C1C |
Safety and Handling
EINECSNumber : 209-463-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only