missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylnaphthalene, 97%

Catalog No. aal0838603
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AAL0838603 1 g
AAL0838606 5 g
2 options

Catalog No. AAL0838603

Supplier: Thermo Scientific Chemicals L0838603

Only null left

CAS: 581-40-8 | C12H12 | 156.228 g/mol

It is used in the identification of individual aromatic hydrocarbons in kerosene fraction (b.p. 150-250°).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the identification of individual aromatic hydrocarbons in kerosene fraction (b.p. 150-250°).

Solubility
Solubility in toluene (almost transparency). Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 581-40-8
Molecular Formula C12H12
Molecular Weight (g/mol) 156.228
MDL Number MFCD00004119
InChI Key WWGUMAYGTYQSGA-UHFFFAOYSA-N
Synonym guajen, naphthalene, 2,3-dimethyl, unii-35dj6se17o, 2,3-dmn, 2,3-dimethyl naphthalene, acmc-1ao3c, 2,3-dimethylnaphthalene, naphthalene, 2,3-dimethyl-8ci 9ci
PubChem CID 11386
ChEBI CHEBI:48615
IUPAC Name 2,3-dimethylnaphthalene
SMILES CC1=CC2=CC=CC=C2C=C1C

Specifications

Melting Point 103°C to 106°C
Boiling Point 265°C to 268°C
Quantity 1 g
Beilstein 1852621
Formula Weight 156.23
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylnaphthalene
EINECSNumber 209-463-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only