missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylnaphthalene, 97%, Thermo Scientific™

Chemical Identifiers

CAS 581-40-8
Molecular Formula C12H12
Molecular Weight (g/mol) 156.23
InChI Key WWGUMAYGTYQSGA-UHFFFAOYSA-N
Synonym guajen, naphthalene, 2,3-dimethyl, unii-35dj6se17o, 2,3-dmn, 2,3-dimethyl naphthalene, acmc-1ao3c, 2,3-dimethylnaphthalene, naphthalene, 2,3-dimethyl-8ci 9ci
PubChem CID 11386
ChEBI CHEBI:48615
IUPAC Name 2,3-dimethylnaphthalene
SMILES CC1=CC2=CC=CC=C2C=C1C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC116450010
SDS View Documents
Thermo Scientific Chemicals
116450010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC116450050
SDS View Documents
Thermo Scientific Chemicals
116450050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

581-40-8
156.23
guajen, naphthalene, 2,3-dimethyl, unii-35dj6se17o, 2,3-dmn, 2,3-dimethyl naphthalene, acmc-1ao3c, 2,3-dimethylnaphthalene, naphthalene, 2,3-dimethyl-8ci 9ci
CHEBI:48615
CC1=CC2=CC=CC=C2C=C1C
C12H12
WWGUMAYGTYQSGA-UHFFFAOYSA-N
11386
2,3-dimethylnaphthalene

Specifications

581-40-8
Authentic
Glass Bottle
C10H6(CH3)2
05,IV,1713
Solubility in water: insoluble.
CC1=CC2=CC=CC=C2C=C1C
156.23
CHEBI:48615
97%
269.0°C
96% min. (GC)
C12H12
1g
guajen, naphthalene, 2,3-dimethyl, unii-35dj6se17o, 2,3-dmn, 2,3-dimethyl naphthalene, acmc-1ao3c, 2,3-dimethylnaphthalene, naphthalene, 2,3-dimethyl-8ci 9ci
WWGUMAYGTYQSGA-UHFFFAOYSA-N
2,3-dimethylnaphthalene
11386
156.23
2,3-Dimethylnaphthalene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 209-463-5

SDS
Documents

Documents

RUO – Research Use Only