missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylnaphthalene, 97%

Catalog No. AC116450010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC116450010 1 g
AC116450050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC116450010 Supplier Thermo Scientific Chemicals Supplier No. 116450010

CAS: 581-40-8 | C12H12 | 156.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 581-40-8
Molecular Formula C12H12
Molecular Weight (g/mol) 156.23
InChI Key WWGUMAYGTYQSGA-UHFFFAOYSA-N
Synonym guajen, naphthalene, 2,3-dimethyl, unii-35dj6se17o, 2,3-dmn, 2,3-dimethyl naphthalene, acmc-1ao3c, 2,3-dimethylnaphthalene, naphthalene, 2,3-dimethyl-8ci 9ci
PubChem CID 11386
ChEBI CHEBI:48615
IUPAC Name 2,3-dimethylnaphthalene
SMILES CC1=CC2=CC=CC=C2C=C1C

Specifications

Melting Point 102.0°C to 104.0°C
Boiling Point 269.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula C10H6(CH3)2
Quantity 1 g
Beilstein 05,IV,1713
Solubility Information Solubility in water: insoluble.
Formula Weight 156.23
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylnaphthalene
Show More Show Less
EINECSNumber 209-463-5

RUO – Research Use Only