missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylhexane, Thermo Scientific™

Catalog No. AC469270050
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469270050 5 mL
AC469270010 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC469270050 Supplier Thermo Scientific Chemicals Supplier No. 469270050

Chemical Identifiers

CAS 584-94-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number 00039951
InChI Key JXPOLSKBTUYKJB-UHFFFAOYNA-N
IUPAC Name 2,3-dimethylhexane
SMILES CCCC(C)C(C)C

Specifications

Alpha Vector DIMETHYLHEXANE
Color Clear colorless to almost colorless
Flash Point 7°C
Infrared Spectrum Conforms
Packaging Glass Bottle
Refractive Index 1.3990 to 1.4010 (20°C, 589 nm)
Quantity 5 mL
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylhexane
Hazard Category Flame
Hazard Statement Exclamation mark
EINECSNumber 209-547-1
Recommended Storage Normal conditions