missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylhexane 94.0+%, TCI America™

Catalog No. D12255ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12255ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12255ML Supplier TCI America Supplier No. D12255ML

Chemical Identifiers

CAS 584-94-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number MFCD00039951
InChI Key JXPOLSKBTUYKJB-UHFFFAOYNA-N
PubChem CID 11447
IUPAC Name 2,3-dimethylhexane
SMILES CCCC(C)C(C)C

Specifications

Color Colorless
Boiling Point 115°C
Quantity 5 mL
UN Number 1262
Formula Weight 114.23
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylhexane
EINECSNumber (2)-0008
TSCA No