missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylheptane 99.0+%, TCI America™

Supplier:  TCI America D12031ML

Catalog No. D12031ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,3-Dimethylheptane
Colorless
C9H20
1 mL
WBRFDUJXCLCKPX-UHFFFAOYSA-N
2,3-dimethylheptane
26375
≥99.0% (GC)
3074-71-3
140°C
MFCD00048819
1920
CCCCC(C)C(C)C
128.259
128.26
Liquid

Chemical Identifiers

3074-71-3
128.259
WBRFDUJXCLCKPX-UHFFFAOYSA-N
2,3-dimethylheptane
C9H20
MFCD00048819
26375
CCCCC(C)C(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0009

TSCA : No

SDS
Documents

Documents

Promotions

Promotions