missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethylfuran 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| UN Number | 1993 |
| Formula Weight | 96.13 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,3-Dimethylfuran |
Safety and Handling
| TSCA | No |