missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 2,3-Dimethylbutane

Catalog No. D151602500M
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D151602500M 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D151602500M Supplier Sigma Aldrich Supplier No. D151602500ML

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,3-Dimethylbutane

Specifications

CAS 79-29-8
Melting Point -129°C (lit.)
Density 0.662 g/mL (at 25°C (literature))
Boiling Point 58°C (lit.)
Molecular Formula C6H14
Refractive Index n20/D 1.375 (literature)
Linear Formula (CH3)2 CHCH(CH3)2
Quantity 500 mL
MDL Number MFCD00008925
Synonym Diisopropyl
Molecular Weight (g/mol) 86.18
Percent Purity 98%
Show More Show Less