missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-2-butene 96.0+%, TCI America™

Catalog No. p-7126378
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D069325ML 25 mL
D0693100ML 100 mL
D0693500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D069325ML Supplier TCI America Supplier No. D069325ML

Chemical Identifiers

CAS 563-79-1
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00008897
InChI Key WGLLSSPDPJPLOR-UHFFFAOYSA-N
Synonym 2,3-dimethyl-2-butene, tetramethylethylene, 2-butene, 2,3-dimethyl, 2,3-dimethylbutene-2, 1,1,2,2-tetramethylethylene, tetramethylethene, 2,3-dimethyl-but-2-ene, ch3 2c=c ch3 2, 2-butene,3-dimethyl, acmc-1akps
PubChem CID 11250
IUPAC Name 2,3-dimethylbut-2-ene
SMILES CC(=C(C)C)C

Specifications

Color Yellow
Boiling Point 73°C
Quantity 25 mL
UN Number 3295
Formula Weight 84.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dimethyl-2-butene
EINECSNumber (2)-0031
TSCA Yes