missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-2,3-butanediamine Dihydrochloride 98.0+%, TCI America™

Catalog No. D32121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32121G 1 g
D32125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32121G Supplier TCI America Supplier No. D32121G

Chemical Identifiers

CAS 75804-28-3
Molecular Formula C6H18Cl2N2
Molecular Weight (g/mol) 189.124
InChI Key BRLVXFONMZRJCD-UHFFFAOYSA-N
Synonym 2,3-Diamino-2,3-dimethylbutane Dihydrochloride, 1,1,2,2-Tetramethylethylenediamine Dihydrochloride
PubChem CID 18760495
IUPAC Name 2,3-dimethylbutane-2,3-diamine;dihydrochloride
SMILES CC(C)(C(C)(C)N)N.Cl.Cl

Specifications

Color White
Quantity 1 g
Formula Weight 189.12
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethyl-2,3-butanediamine Dihydrochloride