missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-1-butene 98.0+%, TCI America™

Catalog No. D1139500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1139500ML 500 mL
D113925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1139500ML Supplier TCI America Supplier No. D1139500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 563-78-0
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00008923
InChI Key OWWIWYDDISJUMY-UHFFFAOYSA-N
Synonym 2,3-dimethyl-1-butene, 1-butene, 2,3-dimethyl, 2,3-dimethylbutene, 2,3-dimethylbutene-1, 2,3-dimethyl butene, acmc-1aqwm, butene, 2,3-dimethyl, ch3 2chc ch3 =ch2, 1-butene, 2,3-dimethyl-8ci 9ci
PubChem CID 11249
IUPAC Name 2,3-dimethylbut-1-ene
SMILES CC(C)C(=C)C

Specifications

Color Colorless
Boiling Point 55°C
Quantity 500 mL
UN Number 3295
Formula Weight 84.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dimethyl-1-butene
EINECSNumber (2)-0031
TSCA No