missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-1-butene, 99%

CAS: 563-78-0 | C6H12 | 84.162 g/mol

Supplier:  Thermo Scientific Chemicals L0567514

 View more versions of this product

Catalog No. AAL0567514

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,3-Dimethyl-1-butene
-157°C
56°C to 57°C
C6H12
MFCD00008923
UN3295
2,3-dimethyl-1-butene, 1-butene, 2,3-dimethyl, 2,3-dimethylbutene, 2,3-dimethylbutene-1, 2,3-dimethyl butene, acmc-1aqwm, butene, 2,3-dimethyl, ch3 2chc ch3 =ch2, 1-butene, 2,3-dimethyl-8ci 9ci
CC(C)C(=C)C
84.162
84.16
563-78-0
0.679
−18°C (−°F)
1.391
25 g
505982
OWWIWYDDISJUMY-UHFFFAOYSA-N
2,3-dimethylbut-1-ene
11249
99%

Chemical Identifiers

563-78-0
84.162
OWWIWYDDISJUMY-UHFFFAOYSA-N
11249
CC(C)C(=C)C
C6H12
MFCD00008923
2,3-dimethyl-1-butene, 1-butene, 2,3-dimethyl, 2,3-dimethylbutene, 2,3-dimethylbutene-1, 2,3-dimethyl butene, acmc-1aqwm, butene, 2,3-dimethyl, ch3 2chc ch3 =ch2, 1-butene, 2,3-dimethyl-8ci 9ci
2,3-dimethylbut-1-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 209-262-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only