missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxybenzeneboronic acid, 97+%

CAS: 40972-86-9 | C8H11BO4 | 181.98 g/mol

$159.93 - $563.14

Chemical Identifiers

CAS 40972-86-9
Molecular Formula C8H11BO4
Molecular Weight (g/mol) 181.98
MDL Number MFCD02683112
InChI Key VREWSCMOGIXMDQ-UHFFFAOYSA-N
Synonym 2,3-dimethoxybenzeneboronic acid, 2,3-dimethoxyphenyl boronic acid, boronic acid, 2,3-dimethoxyphenyl, 2,3-dimethoxyphenylboronicacid, 3-boronoveratrole, pubchem14109, acmc-209jhv, ksc235o1n, 2,3-dimethoxy-phenylboronic acid
PubChem CID 5156491
IUPAC Name (2,3-dimethoxyphenyl)boronic acid
SMILES COC1=CC=CC(B(O)O)=C1OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2412506
View Documents
Thermo Scientific Chemicals
B2412506
5 g
Each for $159.93
Only null left
 
AAB2412514
View Documents
Thermo Scientific Chemicals
B2412514
25 g
Each for $563.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

40972-86-9
181.98
VREWSCMOGIXMDQ-UHFFFAOYSA-N
5156491
COC1=CC=CC(B(O)O)=C1OC
C8H11BO4
MFCD02683112
2,3-dimethoxybenzeneboronic acid, 2,3-dimethoxyphenyl boronic acid, boronic acid, 2,3-dimethoxyphenyl, 2,3-dimethoxyphenylboronicacid, 3-boronoveratrole, pubchem14109, acmc-209jhv, ksc235o1n, 2,3-dimethoxy-phenylboronic acid
(2,3-dimethoxyphenyl)boronic acid

Specifications

40972-86-9
C8H11BO4
5 g
2,3-dimethoxybenzeneboronic acid, 2,3-dimethoxyphenyl boronic acid, boronic acid, 2,3-dimethoxyphenyl, 2,3-dimethoxyphenylboronicacid, 3-boronoveratrole, pubchem14109, acmc-209jhv, ksc235o1n, 2,3-dimethoxy-phenylboronic acid
COC1=CC=CC(B(O)O)=C1OC
181.98
181.98
2,3-Dimethoxybenzeneboronic acid
67°C to 71°C
MFCD02683112
3051295
VREWSCMOGIXMDQ-UHFFFAOYSA-N
(2,3-dimethoxyphenyl)boronic acid
5156491
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only