missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethoxybenzaldehyde 98.0+%, TCI America™
$86.64 - $823.97
Chemical Identifiers
| CAS | 86-51-1 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.176 |
| MDL Number | MFCD00003309 |
| InChI Key | JIVGSHFYXPRRSZ-UHFFFAOYSA-N |
| Synonym | o-veratraldehyde, benzaldehyde, 2,3-dimethoxy, unii-8alp3sy00l, 2,3-dimethoxy-benzaldehyde, 8alp3sy00l, benzaldehyde,3-dimethoxy, dimethoxybenzaldehyde, pubchem8216, benzaldehyde, dimethoxy, upcmld00wstructure66 |
| PubChem CID | 66581 |
| IUPAC Name | 2,3-dimethoxybenzaldehyde |
| SMILES | COC1=CC=CC(=C1OC)C=O |
Chemical Identifiers
| 86-51-1 | |
| 166.176 | |
| JIVGSHFYXPRRSZ-UHFFFAOYSA-N | |
| 66581 | |
| COC1=CC=CC(=C1OC)C=O |
| C9H10O3 | |
| MFCD00003309 | |
| o-veratraldehyde, benzaldehyde, 2,3-dimethoxy, unii-8alp3sy00l, 2,3-dimethoxy-benzaldehyde, 8alp3sy00l, benzaldehyde,3-dimethoxy, dimethoxybenzaldehyde, pubchem8216, benzaldehyde, dimethoxy, upcmld00wstructure66 | |
| 2,3-dimethoxybenzaldehyde |
Specifications
| 86-51-1 | |
| White-Yellow | |
| C9H10O3 | |
| 25 g | |
| JIVGSHFYXPRRSZ-UHFFFAOYSA-N | |
| 2,3-dimethoxybenzaldehyde | |
| 66581 | |
| ≥98.0% (GC) | |
| 2,3-Dimethoxybenzaldehyde |
| 53°C | |
| 256°C | |
| MFCD00003309 | |
| o-veratraldehyde, benzaldehyde, 2,3-dimethoxy, unii-8alp3sy00l, 2,3-dimethoxy-benzaldehyde, 8alp3sy00l, benzaldehyde,3-dimethoxy, dimethoxybenzaldehyde, pubchem8216, benzaldehyde, dimethoxy, upcmld00wstructure66 | |
| COC1=CC=CC(=C1OC)C=O | |
| 166.176 | |
| 166.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2665
TSCA : No