missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxybenzaldehyde, 98+%

CAS: 86-51-1 | C9H10O3 | 166.176 g/mol

$84.38 - $284.22

Chemical Identifiers

CAS 86-51-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00003309
InChI Key JIVGSHFYXPRRSZ-UHFFFAOYSA-N
Synonym o-veratraldehyde, benzaldehyde, 2,3-dimethoxy, unii-8alp3sy00l, 2,3-dimethoxy-benzaldehyde, 8alp3sy00l, benzaldehyde,3-dimethoxy, dimethoxybenzaldehyde, pubchem8216, benzaldehyde, dimethoxy, upcmld00wstructure66
PubChem CID 66581
IUPAC Name 2,3-dimethoxybenzaldehyde
SMILES COC1=CC=CC(=C1OC)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1408014
View Documents
Thermo Scientific Chemicals
A1408014
25 g
Each for $84.38
Only null left
 
AAA1408022
View Documents
Thermo Scientific Chemicals
A1408022
100 g
Each for $284.22
Only null left
 
Description

Description

2,3-Dimethoxybenzaldehyde possess antifungal activity of redox-active benzaldehydes that target cellular antioxidation. It could function as chemosensitizing agents in concert with conventional drugs to improve antifungal efficacy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethoxybenzaldehyde possess antifungal activity of redox-active benzaldehydes that target cellular antioxidation. It could function as chemosensitizing agents in concert with conventional drugs to improve antifungal efficacy.

Solubility
Solubility in methanol with almost transparency.

Notes
Air sensitive. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

86-51-1
166.176
JIVGSHFYXPRRSZ-UHFFFAOYSA-N
66581
COC1=CC=CC(=C1OC)C=O
C9H10O3
MFCD00003309
o-veratraldehyde, benzaldehyde, 2,3-dimethoxy, unii-8alp3sy00l, 2,3-dimethoxy-benzaldehyde, 8alp3sy00l, benzaldehyde,3-dimethoxy, dimethoxybenzaldehyde, pubchem8216, benzaldehyde, dimethoxy, upcmld00wstructure66
2,3-dimethoxybenzaldehyde

Specifications

86-51-1
178°C (20 mmHg)
C9H10O3
25 g
Air Sensitive
JIVGSHFYXPRRSZ-UHFFFAOYSA-N
2,3-dimethoxybenzaldehyde
66581
≥98%
51°C to 52°C
>110°C (230°F)
MFCD00003309
908264
o-veratraldehyde, benzaldehyde, 2,3-dimethoxy, unii-8alp3sy00l, 2,3-dimethoxy-benzaldehyde, 8alp3sy00l, benzaldehyde,3-dimethoxy, dimethoxybenzaldehyde, pubchem8216, benzaldehyde, dimethoxy, upcmld00wstructure66
COC1=CC=CC(=C1OC)C=O
166.176
166.18
2,3-Dimethoxybenzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 201-677-7

RTECSNumber : CU5732000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only