missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimercaptopropanol, 97%

Catalog No. AAL0395306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0395306 5 g
AAL0395303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0395306 Supplier Thermo Scientific Chemicals Supplier No. L0395306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 59-52-9 | C3H8OS2 | 124.216 g/mol

British Anti-Lewisite. A chelator that inhibits sugar nucleotide degradation.2,3-Dimercaptopropanol is used as a chelating agent involved in the treatment of arsenic, antimony, lead, gold and mercury poisoning. It is also used as an antidote to the warfare agent Lewisite. Further, it is used for the treatment of Wilson's disease. In addition to this, it is used as an adjunct in the treatment of the acute encephalopathy of lead toxicity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water, ether, ethanol, vegetable oils and acetone. Slightly miscible with chloroform.

Notes
Air sensitive. Store in a cool place. Incompatible with bases, oxidizing agents, reducing agents and alkali metals.

Chemical Identifiers

CAS 59-52-9
Molecular Formula C3H8OS2
Molecular Weight (g/mol) 124.216
MDL Number MFCD00004864
InChI Key WQABCVAJNWAXTE-UHFFFAOYSA-N
Synonym dimercaprol, 2,3-dimercapto-1-propanol, 2,3-dimercaptopropanol, dithioglycerine, dicaptol, sulfactin, dimercaptopropanol, british anti-lewisite, 2,3-dithiopropanol, antoxol
PubChem CID 3080
ChEBI CHEBI:64198
IUPAC Name 2,3-bis(sulfanyl)propan-1-ol
SMILES C(C(CS)S)O

Specifications

Density 1.244
Boiling Point 120°C (15 mmHg)
Flash Point >110°C (230°F)
Odor Garlic-like
Refractive Index 1.574
Quantity 5 g
UN Number UN2810
Beilstein 1732058
Sensitivity Air Sensitive
Merck Index 14,3209
Solubility Information Miscible with water,ether,ethanol,vegetable oils and acetone. Slightly miscible with chloroform.
Formula Weight 124.22
Percent Purity 97%
Chemical Name or Material 2,3-Dimercaptopropanol
Show More Show Less
Hazard Category H301-H315-H317-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H317
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 200-433-7
RTECSNumber UB2625000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only