missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimercapto-1-propanol 95.0+%, TCI America™

Catalog No. D06211G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06211G 1 g
D06215G 5 g
D062125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D06211G Supplier TCI America Supplier No. D06211G

Chemical Identifiers

CAS 59-52-9
Molecular Formula C3H8OS2
Molecular Weight (g/mol) 124.216
MDL Number MFCD00004864
InChI Key WQABCVAJNWAXTE-UHFFFAOYSA-N
Synonym dimercaprol, 2,3-dimercapto-1-propanol, 2,3-dimercaptopropanol, dithioglycerine, dicaptol, sulfactin, dimercaptopropanol, british anti-lewisite, 2,3-dithiopropanol, antoxol
PubChem CID 3080
ChEBI CHEBI:64198
IUPAC Name 2,3-bis(sulfanyl)propan-1-ol
SMILES C(C(CS)S)O

Specifications

Color Yellow
Boiling Point 120°C
Quantity 1 g
UN Number 2810
Formula Weight 124.22
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,3-Dimercapto-1-propanol
RTECSNumber UB2625000
TSCA Yes