missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydroxypyridine 99.0+%, TCI America™

Catalog No. P07555G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P07555G Supplier TCI America Supplier No. P07555G
Only null left

Chemical Identifiers

CAS 16867-04-2
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00006271
InChI Key GGOZGYRTNQBSSA-UHFFFAOYSA-N
Synonym 2,3-dihydroxypyridine, 2,3-pyridinediol, pyridine-2,3-diol, 3-hydroxy-2-pyridone, 2 1h-pyridinone, 3-hydroxy, 3-hydroxy-2 1h-pyridinone, 3-hydroxypyridin-2 1h-one, 2 1h-pyridone, 3-hydroxy, hydroxypyridone, unii-7hy4bcj130
PubChem CID 28115
IUPAC Name 3-hydroxy-1H-pyridin-2-one
SMILES C1=CNC(=O)C(=C1)O

Specifications

Quantity 5 g
Formula Weight 111.10
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dihydroxypyridine
RTECSNumber UV1146700
TSCA Yes