missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydroxybenzaldehyde 98.0+%, TCI America™
$158.44 - $549.89
Chemical Identifiers
| CAS | 24677-78-9 |
|---|---|
| Molecular Formula | C7H6O3 |
| Molecular Weight (g/mol) | 138.122 |
| MDL Number | MFCD00003324 |
| InChI Key | IXWOUPGDGMCKGT-UHFFFAOYSA-N |
| Synonym | o-pyrocatechualdehyde, 5,6-dihydroxybenzaldehyde, benzaldehyde, 2,3-dihydroxy, benzaldehyde, dihydroxy, 2,3-dihydroxybenzaldehyde, dihydroxybenzaldehyde, pubchem6561, acmc-1clux, 2,3-dihydroxy benzaldehyde, 2,3-dihydroxy-benzaldehyde |
| PubChem CID | 90579 |
| ChEBI | CHEBI:50197 |
| IUPAC Name | 2,3-dihydroxybenzaldehyde |
| SMILES | C1=CC(=C(C(=C1)O)O)C=O |
Chemical Identifiers
| 24677-78-9 | |
| 138.122 | |
| IXWOUPGDGMCKGT-UHFFFAOYSA-N | |
| 90579 | |
| 2,3-dihydroxybenzaldehyde |
| C7H6O3 | |
| MFCD00003324 | |
| o-pyrocatechualdehyde, 5,6-dihydroxybenzaldehyde, benzaldehyde, 2,3-dihydroxy, benzaldehyde, dihydroxy, 2,3-dihydroxybenzaldehyde, dihydroxybenzaldehyde, pubchem6561, acmc-1clux, 2,3-dihydroxy benzaldehyde, 2,3-dihydroxy-benzaldehyde | |
| CHEBI:50197 | |
| C1=CC(=C(C(=C1)O)O)C=O |
Specifications
| 24677-78-9 | |
| 120°C | |
| MFCD00003324 | |
| o-pyrocatechualdehyde, 5,6-dihydroxybenzaldehyde, benzaldehyde, 2,3-dihydroxy, benzaldehyde, dihydroxy, 2,3-dihydroxybenzaldehyde, dihydroxybenzaldehyde, pubchem6561, acmc-1clux, 2,3-dihydroxy benzaldehyde, 2,3-dihydroxy-benzaldehyde | |
| C1=CC(=C(C(=C1)O)O)C=O | |
| 138.122 | |
| CHEBI:50197 | |
| ≥98.0% (GC,T) | |
| 2,3-Dihydroxybenzaldehyde |
| 107°C | |
| C7H6O3 | |
| 5 g | |
| IXWOUPGDGMCKGT-UHFFFAOYSA-N | |
| 2,3-dihydroxybenzaldehyde | |
| 90579 | |
| 138.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No