missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydroxybenzaldehyde 98.0+%, TCI America™

Catalog No. D14785G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14785G 5 g
D147825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D14785G Supplier TCI America Supplier No. D14785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24677-78-9
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.122
MDL Number MFCD00003324
InChI Key IXWOUPGDGMCKGT-UHFFFAOYSA-N
Synonym o-pyrocatechualdehyde, 5,6-dihydroxybenzaldehyde, benzaldehyde, 2,3-dihydroxy, benzaldehyde, dihydroxy, 2,3-dihydroxybenzaldehyde, dihydroxybenzaldehyde, pubchem6561, acmc-1clux, 2,3-dihydroxy benzaldehyde, 2,3-dihydroxy-benzaldehyde
PubChem CID 90579
ChEBI CHEBI:50197
IUPAC Name 2,3-dihydroxybenzaldehyde
SMILES C1=CC(=C(C(=C1)O)O)C=O

Specifications

Melting Point 107°C
Boiling Point 120°C
Quantity 5 g
Formula Weight 138.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dihydroxybenzaldehyde