missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydroxybenzaldehyde, 97%

Catalog No. AAA1349022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1349022 100 g
AAA1349006 5 g
AAA1349014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1349022 Supplier Thermo Scientific Chemicals Supplier No. A1349022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24677-78-9 | C7H6O3 | 138.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24677-78-9
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.122
MDL Number MFCD00003324
InChI Key IXWOUPGDGMCKGT-UHFFFAOYSA-N
Synonym o-pyrocatechualdehyde, 5,6-dihydroxybenzaldehyde, benzaldehyde, 2,3-dihydroxy, benzaldehyde, dihydroxy, 2,3-dihydroxybenzaldehyde, dihydroxybenzaldehyde, pubchem6561, acmc-1clux, 2,3-dihydroxy benzaldehyde, 2,3-dihydroxy-benzaldehyde
PubChem CID 90579
ChEBI CHEBI:50197
IUPAC Name 2,3-dihydroxybenzaldehyde
SMILES C1=CC(=C(C(=C1)O)O)C=O

Specifications

Melting Point 106°C to 109°C
Quantity 100 g
Beilstein 2041781
Sensitivity Air Sensitive
Formula Weight 138.12
Percent Purity 97%
Chemical Name or Material 2,3-Dihydroxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 246-398-1
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only