missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrofuran 98.0+%, TCI America™

Catalog No. p-7126721
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D141525ML 25 mL
D1415100ML 100 mL
D1415500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D141525ML Supplier TCI America Supplier No. D141525ML
Only null left

Chemical Identifiers

CAS 1191-99-7
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00003205
InChI Key JKTCBAGSMQIFNL-UHFFFAOYSA-N
PubChem CID 70934
ChEBI CHEBI:51662
IUPAC Name 2,3-dihydrofuran
SMILES C1CC=CO1

Specifications

Color Colorless
Boiling Point 54°C
Quantity 25 mL
UN Number 1993
Synonym 4,5-dihydrofuran, furan, 2,3-dihydro, furan, dihydro, dihydrofuran, 2,3 dihydrofuran, dihydrofurane, 2,3dihydrofuran, furan,3-dihydro, 2,3-dihydrofurane, 2,3-dihydro-furan
Formula Weight 70.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dihydrofuran
EINECSNumber (9)-0595
TSCA Yes