missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydrobenzofuran 98.0+%, TCI America™
$67.38 - $215.62
Chemical Identifiers
| CAS | 496-16-2 |
|---|---|
| Molecular Formula | C8H8O |
| Molecular Weight (g/mol) | 120.151 |
| MDL Number | MFCD00005855 |
| InChI Key | HBEDSQVIWPRPAY-UHFFFAOYSA-N |
| Synonym | 2,3-dihydrobenzofuran, coumaran, 2,3-dihydrobenzo b furan, benzofuran, 2,3-dihydro, dihydrobenzofuran, kumaran, dihydrocoumarone, 2,3-dihydro-benzofuran, 2,3-dihydrobenzofurane |
| PubChem CID | 10329 |
| ChEBI | CHEBI:87607 |
| IUPAC Name | 2,3-dihydro-1-benzofuran |
| SMILES | C1COC2=CC=CC=C21 |
Chemical Identifiers
| 496-16-2 | |
| 120.151 | |
| HBEDSQVIWPRPAY-UHFFFAOYSA-N | |
| 10329 | |
| 2,3-dihydro-1-benzofuran |
| C8H8O | |
| MFCD00005855 | |
| 2,3-dihydrobenzofuran, coumaran, 2,3-dihydrobenzo b furan, benzofuran, 2,3-dihydro, dihydrobenzofuran, kumaran, dihydrocoumarone, 2,3-dihydro-benzofuran, 2,3-dihydrobenzofurane | |
| CHEBI:87607 | |
| C1COC2=CC=CC=C21 |
Specifications
| 496-16-2 | |
| 189°C | |
| MFCD00005855 | |
| 2,3-dihydrobenzofuran, coumaran, 2,3-dihydrobenzo b furan, benzofuran, 2,3-dihydro, dihydrobenzofuran, kumaran, dihydrocoumarone, 2,3-dihydro-benzofuran, 2,3-dihydrobenzofurane | |
| C1COC2=CC=CC=C21 | |
| 120.151 | |
| CHEBI:87607 | |
| ≥98.0% (GC) | |
| 2,3-Dihydrobenzofuran |
| Yellow | |
| C8H8O | |
| 5 g | |
| HBEDSQVIWPRPAY-UHFFFAOYSA-N | |
| 2,3-dihydro-1-benzofuran | |
| 10329 | |
| 120.15 | |
| Liquid |
Safety and Handling
EINECSNumber : (5)-6777
TSCA : Yes