missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzofuran 98.0+%, TCI America™

Catalog No. D15835G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15835G 5 g
D158325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15835G Supplier TCI America Supplier No. D15835G
Only null left

Chemical Identifiers

CAS 496-16-2
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00005855
InChI Key HBEDSQVIWPRPAY-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran, coumaran, 2,3-dihydrobenzo b furan, benzofuran, 2,3-dihydro, dihydrobenzofuran, kumaran, dihydrocoumarone, 2,3-dihydro-benzofuran, 2,3-dihydrobenzofurane
PubChem CID 10329
ChEBI CHEBI:87607
IUPAC Name 2,3-dihydro-1-benzofuran
SMILES C1COC2=CC=CC=C21

Specifications

Color Yellow
Boiling Point 189°C
Quantity 5 g
Formula Weight 120.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dihydrobenzofuran
EINECSNumber (5)-6777
TSCA Yes