missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzofuran-5-sulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7127786
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D35531G 1 g
D35535G 5 g
2 options

Catalog No. D35531G

Supplier: TCI America D35531G

Only null left

Chemical Identifiers

CAS 115010-11-2
Molecular Formula C8H7ClO3S
Molecular Weight (g/mol) 218.65
MDL Number MFCD02677703
InChI Key RVWYPBARHGPULM-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-sulfonyl chloride, 2,3-dihydro-1-benzofuran-5-sulfonoylchloride, 2,3-dihydro-1-benzofuran-5-sulfonoyl chloride, 2,3-dihydrobenzofuran-5-sulfonylchloride, 2,3-dihydro-benzofuran-5-sulfonyl chloride, 2,3-dihydrobenzo b furan-5-sulfonyl chloride, pubchem14874, 5-benzofuransulfonylchloride, 2,3-dihydro, 2,3-dihydro-1-benzofuran-5-sulfonylchloride, chloro-2,3-dihydrobenzo b furan-5-ylsulfone
PubChem CID 2776154
IUPAC Name 2,3-dihydro-1-benzofuran-5-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC2=C(OCC2)C=C1

Specifications

Melting Point 85°C
Quantity 1 g
UN Number 3261
Formula Weight 218.65
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dihydrobenzofuran-5-sulfonyl Chloride
TSCA No
Recommended Storage Refrigerator