missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzofuran-5-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D4711200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4711200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4711200MG Supplier TCI America Supplier No. D4711200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 227305-69-3
Molecular Formula C8H9BO3
Molecular Weight (g/mol) 163.97
MDL Number MFCD02681979
InChI Key ZIXLJHSFAMVHPC-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-boronic acid, 2,3-dihydrobenzofuran-5-ylboronic acid, 2,3-dihydrobenzofuran-5-yl boronic acid, 2,3-dihydrobenzo b furan-5-boronic acid, 2,3-dihydrobenzofuran-5-yl-5-boronic acid, boronic acid, 2,3-dihydro-5-benzofuranyl, 2,3-dihydro-1-benzofuran-5-yl-boranediol, 2,3-dihydro-1-benzofuran-5-yl boronic acid, pubchem7848, zlchem 1108
PubChem CID 2773380
IUPAC Name (2,3-dihydro-1-benzofuran-5-yl)boronic acid
SMILES OB(O)C1=CC=C2OCCC2=C1

Specifications

Melting Point 240°C
Color White-Yellow
Quantity 200 mg
Formula Weight 163.97
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dihydrobenzofuran-5-boronic Acid (contains varying amounts of Anhydride)