missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzo[b]furan-5-sulfonyl chloride, 97%

Catalog No. AAH33579MD
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33579MD 250 mg
AAH3357906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33579MD Supplier Thermo Scientific Chemicals Supplier No. H33579MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 115010-11-2 | C8H7ClO3S | 218.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115010-11-2
Molecular Formula C8H7ClO3S
Molecular Weight (g/mol) 218.65
MDL Number MFCD02677703
InChI Key RVWYPBARHGPULM-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-sulfonyl chloride, 2,3-dihydro-1-benzofuran-5-sulfonoylchloride, 2,3-dihydro-1-benzofuran-5-sulfonoyl chloride, 2,3-dihydrobenzofuran-5-sulfonylchloride, 2,3-dihydro-benzofuran-5-sulfonyl chloride, 2,3-dihydrobenzo b furan-5-sulfonyl chloride, pubchem14874, 5-benzofuransulfonylchloride, 2,3-dihydro, 2,3-dihydro-1-benzofuran-5-sulfonylchloride, chloro-2,3-dihydrobenzo b furan-5-ylsulfone
PubChem CID 2776154
IUPAC Name 2,3-dihydro-1-benzofuran-5-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC2=C(OCC2)C=C1

Specifications

Melting Point 83°C to 85°C
Quantity 250 mg
UN Number UN3261
Formula Weight 218.66
Percent Purity 97%
Chemical Name or Material 2,3-Dihydrobenzo[b]furan-5-sulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only